Assay Details
| Gene Symbol: Pctp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008796.2 | NP_032822.2 | 2 - 3 | 308 | 128 | |
| GenBank mRNA | AF151639.1 | - | 2 - 3 | 279 | 128 | |
| BB614866.1 | - | 156 | 128 | |||
| BC053478.1 | - | 263 | 128 | |||
| BC071234.1 | - | 2 - 3 | 264 | 128 | ||
| BY144797.1 | - | 2 - 3 | 309 | 128 | ||
| Z50024.1 | - | 2 - 3 | 137 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pctp
Genomic Map