Assay Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008818.2 | NP_032844.2 | 195 | 95 | ||
| GenBank mRNA | AK146379.1 | - | 95 | |||
| AK146393.1 | - | 95 | ||||
| AK146449.1 | - | 201 | 95 | |||
| BC056204.1 | - | 2 - 3 | 203 | 30022528 | 95 | |
| DQ058642.1 | - | 1 - 2 | 179 | 95 | ||
| M32484.1 | - | 1 - 2 | 179 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rhox5
Genomic Map