Assay Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008922.2 | NP_032948.1 | 1 - 2 | 167 | 76 | |
| GenBank mRNA | AK076095.1 | - | 1 - 2 | 149 | 76 | |
| AK077760.1 | - | 1 - 2 | 154 | 76 | ||
| AK153970.1 | - | 1 - 2 | 128 | 76 | ||
| AK168246.1 | - | 1 - 2 | 160 | 76 | ||
| BC019500.1 | - | 1 - 2 | 128 | 3602929 | 76 | |
| BY131806.1 | - | 1 - 2 | 168 | 76 | ||
| D13545.1 | - | 1 - 2 | 131 | 76 | ||
| D17385.1 | - | 1 - 2 | 111 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK076095.1,AK077760.1,AK153970.1,AK168246.1,BC019500.1,D13545.1,D17385.1,NM_008922.2 transcripts.
Set Membership: |
|

Prim2
Genomic Map