Assay Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008922.2 | NP_032948.1 | 4 - 5 | 519 | 93 | |
| GenBank mRNA | AK076095.1 | - | 4 - 5 | 501 | 93 | |
| AK077760.1 | - | 4 - 5 | 506 | 93 | ||
| AK153970.1 | - | 4 - 5 | 480 | 93 | ||
| AK168246.1 | - | 4 - 5 | 512 | 93 | ||
| BC019500.1 | - | 4 - 5 | 480 | 3602929 | 93 | |
| D13545.1 | - | 4 - 5 | 483 | 93 | ||
| D17385.1 | - | 4 - 5 | 93 | |||
More Information
Set Membership: |
|

Prim2
Genomic Map