Assay Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008922.2 | NP_032948.1 | 8 - 9 | 940 | 74 | |
| GenBank mRNA | AK076095.1 | - | 8 - 9 | 922 | 74 | |
| AK077760.1 | - | 8 - 9 | 927 | 74 | ||
| AK153970.1 | - | 8 - 9 | 901 | 74 | ||
| AK166687.1 | - | 2 - 3 | 146 | 74 | ||
| AK168246.1 | - | 8 - 9 | 933 | 74 | ||
| BC019500.1 | - | 8 - 9 | 901 | 3602929 | 74 | |
| D13545.1 | - | 8 - 9 | 904 | 74 | ||
| D17385.1 | - | 8 - 9 | 884 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim2
Genomic Map