Assay Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008948.2 | NP_032974.2 | 7 - 8 | 971 | 73 | |
| GenBank mRNA | AK002542.1 | - | 7 - 8 | 692 | 73 | |
| AK083043.1 | - | 933 | 73 | |||
| AK150189.1 | - | 7 - 8 | 907 | 73 | ||
| AK151370.1 | - | 7 - 8 | 691 | 73 | ||
| AK152018.1 | - | 7 - 8 | 710 | 73 | ||
| AK167002.1 | - | 7 - 8 | 919 | 73 | ||
| AK168263.1 | - | 6 - 7 | 640 | 73 | ||
| AK168616.1 | - | 7 - 8 | 691 | 73 | ||
| BC005783.1 | - | 7 - 8 | 860 | 3603053 | 73 | |
| D49686.1 | - | 7 - 8 | 915 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc3
Genomic Map