Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008960.2 | NP_032986.1 | 4 - 5 | 1125 | 73 | |
| GenBank mRNA | AK076980.1 | - | 4 - 5 | 1083 | 73 | |
| AK088717.1 | - | 4 - 5 | 1104 | 73 | ||
| AK148736.1 | - | 4 - 5 | 960 | 73 | ||
| AK164302.1 | - | 4 - 5 | 1108 | 73 | ||
| BC021445.1 | - | 4 - 5 | 474 | 5038272 | 73 | |
| U92437.1 | - | 4 - 5 | 1205 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pten
Genomic Map