| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100035007.mm.1 |
NM_008986.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035007.mm.1 |
NM_008986.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035007.mm.1 |
TC1100003791.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035010.mm.1 |
NM_008986.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035010.mm.1 |
NM_008986.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035010.mm.1 |
TC1100003791.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035012.mm.1 |
NM_008986.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035012.mm.1 |
NM_008986.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035012.mm.1 |
TC1100003791.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035006.mm.1 |
NM_008986.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035006.mm.1 |
NM_008986.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035006.mm.1 |
TC1100003791.mm.1 |
MTA TC |