Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 2 - 3 | 787 | 62 | |
| AK128980.1 | - | 2 - 3 | 863 | 62 | ||
| AK158276.1 | - | 2 - 3 | 811 | 62 | ||
| AK159330.1 | - | 2 - 3 | 791 | 62 | ||
| AK159342.1 | - | 2 - 3 | 810 | 62 | ||
| AK159377.1 | - | 2 - 3 | 781 | 62 | ||
| AK159409.1 | - | 2 - 3 | 811 | 62 | ||
| AK159774.1 | - | 2 - 3 | 782 | 62 | ||
| AK159957.1 | - | 2 - 3 | 782 | 62 | ||
| BC082577.1 | - | 2 - 3 | 799 | 30648530 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map