Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 5 - 6 | 1045 | 151 | |
| AK004712.1 | - | 1 - 2 | 51 | 151 | ||
| AK128980.1 | - | 5 - 6 | 1121 | 151 | ||
| AK158276.1 | - | 5 - 6 | 1069 | 151 | ||
| AK159330.1 | - | 5 - 6 | 1049 | 151 | ||
| AK159342.1 | - | 5 - 6 | 1068 | 151 | ||
| AK159377.1 | - | 5 - 6 | 1039 | 151 | ||
| AK159409.1 | - | 5 - 6 | 1069 | 151 | ||
| AK159774.1 | - | 5 - 6 | 1040 | 151 | ||
| AK159957.1 | - | 5 - 6 | 1040 | 151 | ||
| BC082577.1 | - | 5 - 6 | 1057 | 30648530 | 151 | |
More Information
Set Membership: |
|

Aebp1
Genomic Map