Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 16 - 17 | 2212 | 64 | |
| AK004712.1 | - | 12 - 13 | 1221 | 64 | ||
| AK128980.1 | - | 16 - 17 | 2288 | 64 | ||
| AK158276.1 | - | 16 - 17 | 2239 | 64 | ||
| AK159330.1 | - | 16 - 17 | 2219 | 64 | ||
| AK159342.1 | - | 16 - 17 | 2238 | 64 | ||
| AK159377.1 | - | 16 - 17 | 2209 | 64 | ||
| AK159409.1 | - | 16 - 17 | 2239 | 64 | ||
| AK159774.1 | - | 16 - 17 | 2210 | 64 | ||
| AK159957.1 | - | 16 - 17 | 2210 | 64 | ||
| AK189735.1 | - | 1 - 2 | 99 | 64 | ||
| AK195891.1 | - | 1 - 2 | 64 | |||
| AK195983.1 | - | 1 - 2 | 99 | 64 | ||
| AK212842.1 | - | 1 - 2 | 99 | 64 | ||
| BC082577.1 | - | 16 - 17 | 2224 | 30648530 | 64 | |
| X80478.1 | - | 7 - 8 | 942 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map