Assay Details
| Gene Symbol: Cep131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009734.3 | NP_033864.3 | 8 - 9 | 1081 | 72 | |
| GenBank mRNA | AK133245.1 | - | 8 - 9 | 1083 | 72 | |
| BC141108.1 | - | 8 - 9 | 1021 | 9055720 | 72 | |
| D43921.1 | - | 8 - 9 | 1073 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cep131
Genomic Map