Assay Details
| Gene Symbol: Cep131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009734.3 | NP_033864.3 | 22 - 23 | 2893 | 77 | |
| GenBank mRNA | AK133245.1 | - | 22 - 23 | 2896 | 77 | |
| AK134381.1 | - | 14 - 15 | 2535 | 77 | ||
| BC141108.1 | - | 22 - 23 | 2833 | 9055720 | 77 | |
| D43921.1 | - | 22 - 23 | 2888 | 77 | ||
More Information
Set Membership: |
|

Cep131
Genomic Map