Assay Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037758.2 | NP_001032847.2 | 10 - 11 | 1456 | 70 | |
| NM_001286465.1 | NP_001273394.1 | 10 - 11 | 1456 | 70 | ||
| NM_001286466.1 | NP_001273395.1 | 9 - 10 | 1224 | 70 | ||
| NM_009771.3 | NP_033901.1 | 9 - 10 | 1348 | 70 | ||
| GenBank mRNA | AF081887.1 | - | 9 - 10 | 1333 | 70 | |
| AF099932.1 | - | 9 - 10 | 1243 | 70 | ||
| AF110396.1 | - | 9 - 10 | 1243 | 70 | ||
| AF112979.1 | - | 9 - 10 | 1243 | 70 | ||
| AK032221.1 | - | 10 - 11 | 1419 | 70 | ||
| AK052317.1 | - | 11 - 12 | 1480 | 70 | ||
| AK083257.1 | - | 10 - 11 | 1433 | 70 | ||
| AK145091.1 | - | 9 - 10 | 1160 | 70 | ||
| AK145624.1 | - | 10 - 11 | 1439 | 70 | ||
| AK156660.1 | - | 9 - 10 | 1376 | 70 | ||
| AK220183.1 | - | 10 - 11 | 1454 | 70 | ||
| BC003989.1 | - | 9 - 10 | 1249 | 3491843 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btrc
Genomic Map