Assay Details
| Gene Symbol: Hrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010473.2 | NP_034603.2 | 5 - 6 | 2341 | 80 | |
| GenBank mRNA | AF132218.1 | - | 5 - 6 | 2320 | 80 | |
| AF158597.1 | - | 6 - 7 | 2175 | 80 | ||
| AI322629.1 | - | 5 - 6 | 292 | 80 | ||
| BC141121.1 | - | 7 - 8 | 2160 | 9055733 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994229 | AF158597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994229 | AI322629.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994229 | AF132218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994229 | NM_010473.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670834 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166191 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994229 | AF158597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994229 | AI322629.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994229 | AF132218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994229 | NM_010473.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25092507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166191 | - | - |
| Whole Mouse Genome G4122A | A_51_P267751 | BC141121.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267751 | AF158597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267751 | AI322629.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267751 | AF132218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267751 | NM_010473.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131551 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103149 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134464 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05975390 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05975400 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05975399 | - | - |
| Agilent Mouse cDNA Microarray (G4104A) | 475063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25092449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05975385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05975395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25092485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552926 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419109_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552919 | - | - |
| Mouse Genome 430 2.0 Array | 1419109_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419109_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15464_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15464_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007243.mm.1 | NM_010473.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007243.mm.1 | NM_010473.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007243.mm.1 | TC0700000840.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007242.mm.1 | NM_010473.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007242.mm.1 | NM_010473.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007242.mm.1 | TC0700000840.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007241.mm.1 | NM_010473.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007241.mm.1 | NM_010473.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007241.mm.1 | TC0700000840.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426048_s_at | - | - |
| Mouse Expression Set 430 | 1419109_at | - | - |
| Mouse Expression Set 430 | 1426048_PM_s_at | - | - |
| Mouse Expression Set 430 | 1419109_PM_at | - | - |
| Murine Genome U74v2 Set | 92371_PM_at | - | - |
| Murine Genome U74v2 Set | 92371_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426048_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419109_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419109_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426048_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419109_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419109_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 526036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2690563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002690563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259206 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259206 | - | - |
| mouse-6 v1.1 (Illumina) | scl32687.7.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2690563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32687.7.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259206 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259206 | - | - |

Hrc
Genomic Map