Assay Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010925.2 | NP_035055.2 | 9 - 10 | 1147 | 65 | |
| GenBank mRNA | AF312394.1 | - | 8 - 9 | 910 | 65 | |
| AK077540.1 | - | 9 - 10 | 1072 | 65 | ||
| BC003913.1 | - | 2 - 3 | 243 | 65 | ||
| BC022910.1 | - | 8 - 9 | 1013 | 4165783 | 65 | |
| BC065993.1 | - | 9 - 10 | 1109 | 5684672 | 65 | |
| BC065994.1 | - | 9 - 10 | 1067 | 5685269 | 65 | |
| U79773.1 | - | 10 - 11 | 946 | 65 | ||
| U79774.1 | - | 9 - 10 | 1099 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003913.1 transcripts.
Set Membership: |
|

Rrp1
Genomic Map