Assay Details
| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011059.2 | NP_035189.1 | 1 - 2 | 225 | 66 | |
| GenBank mRNA | AB013848.1 | - | 1 - 2 | 225 | 66 | |
| AK037182.1 | - | 1 - 2 | 226 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Padi1
Genomic Map