Assay Details
| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011059.2 | NP_035189.1 | 12 - 13 | 1581 | 90 | |
| GenBank mRNA | AB013848.1 | - | 12 - 13 | 1581 | 90 | |
| AK037182.1 | - | 12 - 13 | 1582 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi1
Genomic Map