Assay Details
Gene Symbol: Prtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011178.2 | NP_035308.2 | 1 - 2 | 78 | 70 | |
GenBank mRNA | U43525.1 | - | 1 - 2 | 66 | 70 | |
U97073.1 | - | 1 - 2 | 78 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279469 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279471 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279470 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | U97073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | U97073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK075848.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK088582.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK147289.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK150009.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK169286.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK194115.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK199903.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK204960.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | AK220536.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC012042.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC013670.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC025040.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC029143.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC031165.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC049104.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC057354.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | BC061232.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924958 | NM_010956.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641239 | AK051611.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641239 | AK156874.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50876 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175216 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | U97073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | FM210470.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | U43525.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | U97073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | FM210470.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | U97073.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | U97073.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279458 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175216 | - | - |
Whole Mouse Genome G4122A | A_51_P209818 | U97073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | U43525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | U97073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | NM_011178.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | FM210470.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P209818 | U43525.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130501 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175216 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08496356 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279470 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279471 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27279469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08841146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08496356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08841153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08389467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08445250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27279442 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10364532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364530 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364531 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364534 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364533 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419669_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364529 | - | - |
Mouse Genome 430 2.0 Array | 1419669_at | - | - |
Mouse Genome 430 2.0 Array | 1419669_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19152_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19152_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000006007.mm.1 | NM_011178.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006007.mm.1 | NM_011178.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006007.mm.1 | TC1000000822.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006008.mm.1 | NM_011178.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006008.mm.1 | NM_011178.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006008.mm.1 | TC1000000822.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419669_at | - | - |
Mouse Expression Set 430 | 1419669_PM_at | - | - |
Murine Genome U74v2 Set | 104541_at | - | - |
Murine Genome U74v2 Set | 104541_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419669_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419669_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419669_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419669_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 764722 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6900670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900670 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2758029 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2758029 | - | - |
mouse-6 v1.1 (Illumina) | scl38707.5.30_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38707.5.30_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2758029 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2758029 | - | - |
