Assay Details
| Gene Symbol: Pts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011220.2 | NP_035350.1 | 3 - 4 | 188 | 84 | |
| GenBank mRNA | AF043225.1 | - | 3 - 4 | 203 | 84 | |
| AK002614.1 | - | 3 - 4 | 198 | 84 | ||
| AK146987.1 | - | 3 - 4 | 199 | 84 | ||
| BC029013.1 | - | 3 - 4 | 188 | 5357873 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pts
Genomic Map