Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 9 - 10 | 867 | 61 | |
| NM_001286207.1 | NP_001273136.1 | 9 - 10 | 840 | 61 | ||
| NM_011791.3 | NP_035921.2 | 9 - 10 | 957 | 61 | ||
| GenBank mRNA | AB020983.2 | - | 9 - 10 | 939 | 61 | |
| AK087934.1 | - | 9 - 10 | 932 | 61 | ||
| AK132471.1 | - | 8 - 9 | 863 | 61 | ||
| AK145576.1 | - | 9 - 10 | 832 | 61 | ||
| AK145790.1 | - | 9 - 10 | 844 | 61 | ||
| AK145967.1 | - | 9 - 10 | 811 | 61 | ||
| AK146254.1 | - | 9 - 10 | 869 | 61 | ||
| AK146599.1 | - | 9 - 10 | 841 | 61 | ||
| AK146756.1 | - | 9 - 10 | 832 | 61 | ||
| AK146938.1 | - | 9 - 10 | 934 | 61 | ||
| AK183467.1 | - | 4 - 5 | 297 | 61 | ||
| BC012957.1 | - | 9 - 10 | 944 | 4211433 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ash2l
Genomic Map