Assay Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011868.3 | NP_035998.2 | 1 - 2 | 320 | 121 | |
| GenBank mRNA | AF153613.1 | - | 1 - 2 | 292 | 121 | |
| BY251531.1 | - | 1 - 2 | 303 | 121 | ||
| CX242291.1 | - | 1 - 2 | 299 | 121 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF153613.1,NM_011868.3 transcripts.
Set Membership: |
|

Eci2
Genomic Map