Assay Details
| Gene Symbol: Nubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011955.2 | NP_036085.1 | 9 - 10 | 856 | 79 | |
| GenBank mRNA | AF114170.1 | - | 9 - 10 | 833 | 79 | |
| AK020141.1 | - | 4 - 5 | 393 | 79 | ||
| AK157212.1 | - | 9 - 10 | 848 | 79 | ||
| BC031430.1 | - | 9 - 10 | 847 | 79 | ||
| BC047926.1 | - | 7 - 8 | 2031 | 79 | ||
| BC055436.1 | - | 9 - 10 | 844 | 3660324 | 79 | |
| BU614991.1 | - | 2 - 3 | 137 | 79 | ||
| CX210766.1 | - | 6 - 7 | 544 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nubp1
Genomic Map