Assay Details
| Gene Symbol: Nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011956.3 | NP_036086.1 | 1 - 2 | 72 | 70 | |
| GenBank mRNA | AF114169.1 | - | 1 - 2 | 50 | 70 | |
| AK003816.1 | - | 1 - 2 | 70 | 70 | ||
| BC012635.1 | - | 1 - 2 | 73 | 4038123 | 70 | |
| BI735707.1 | - | 1 - 2 | 72 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF114169.1,AK003816.1,BC012635.1,NM_011956.3 transcripts.
Set Membership: |
|

Nubp2
Genomic Map