Assay Details
| Gene Symbol: Nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011956.3 | NP_036086.1 | 5 - 6 | 656 | 65 | |
| GenBank mRNA | AF114169.1 | - | 5 - 6 | 634 | 65 | |
| AK003816.1 | - | 5 - 6 | 654 | 65 | ||
| AK169896.1 | - | 5 - 6 | 613 | 65 | ||
| BC012635.1 | - | 5 - 6 | 657 | 4038123 | 65 | |
| BI735707.1 | - | 5 - 6 | 656 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100087 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100083 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103921 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106964 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31100054 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106964 | - | - |
| Whole Mouse Genome G4122A | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490171 | AK003816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490171 | NM_011956.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490171 | AF114169.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120967 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106964 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100087 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31100083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31100155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13531979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13531995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13531972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13531967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31100050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13462792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10448753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448750 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448752 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416512_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459842_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448748 | - | - |
| Mouse Genome 430 2.0 Array | 1416512_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459842_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1459842_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416512_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26426_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26426_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014740.mm.1 | NM_011956.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014740.mm.1 | NM_011956.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014740.mm.1 | TC1700001646.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014739.mm.1 | NM_011956.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014739.mm.1 | NM_011956.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014739.mm.1 | TC1700001646.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416512_PM_at | - | - |
| Mouse Expression Set 430 | 1416512_at | - | - |
| Mouse Expression Set 430 | 1459842_x_at | - | - |
| Mouse Expression Set 430 | 1459842_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416512_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416512_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416512_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416512_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459842_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459842_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 447954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 730100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3870736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 730100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000730100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003870736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2728440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2723438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2728440 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001628.1_55-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50196.7.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3870736 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 730100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001628.1_55-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50196.7.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730100 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2925570 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2925567 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2723438 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2925570 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2925567 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723438 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50196.7.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001628.1_55-S | - | - |

Nubp2
Genomic Map