Assay Details
| Gene Symbol: Psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011969.2 | NP_036099.1 | 1 - 2 | 192 | 100 | |
| GenBank mRNA | AF019662.1 | - | 1 - 2 | 130 | 100 | |
| AK088615.1 | - | 1 - 2 | 137 | 100 | ||
| AK146765.1 | - | 1 - 2 | 149 | 100 | ||
| AK165563.1 | - | 1 - 2 | 145 | 100 | ||
| AK168234.1 | - | 1 - 2 | 163 | 101 | ||
| BC008222.1 | - | 1 - 2 | 125 | 2581897 | 100 | |
| BY779773.1 | - | 1 - 2 | 180 | 100 | ||
| DV049235.1 | - | 1 - 2 | 161 | 100 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK168234.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
ubiquitin-dependent protein catabolic process
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
identical protein binding
proteolysis
ubiquitin-dependent protein catabolic process
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901047 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901044 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | AK008832.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | AK030337.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | BC036156.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | BC036160.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | BC100511.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | BC117023.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | BC117027.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914100 | NM_001099917.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488513 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134239 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901037 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04807963 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134239 | - | - |
| Whole Mouse Genome G4122A | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | BC014752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | BC008222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | NM_011969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | AF019662.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233603 | AK088615.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112480 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01509075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01509036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901047 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21900967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01962605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01631265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01481621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01481622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01631264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01917019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01479758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21901009 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10490375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490377 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490374 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423567_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423568_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10490370 | - | - |
| Mouse Genome 430 2.0 Array | 1423568_at | - | - |
| Mouse Genome 430 2.0 Array | 1423567_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423568_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423567_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26444_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26444_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200045888.mm.1 | NM_001289476.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045888.mm.1 | NM_011969.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045888.mm.1 | TC0200005399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045885.mm.1 | NM_001289476.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045885.mm.1 | NM_011969.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045885.mm.1 | TC0200005399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045892.mm.1 | NM_001289476.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045892.mm.1 | NM_011969.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045892.mm.1 | TC0200005399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045893.mm.1 | NM_001289476.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045893.mm.1 | NM_011969.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045893.mm.1 | TC0200005399.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423567_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423568_PM_at | - | - |
| Mouse Expression Set 430 | 1423567_a_at | - | - |
| Mouse Expression Set 430 | 1423568_at | - | - |
| Murine Genome U74v2 Set | 93988_PM_at | - | - |
| Murine Genome U74v2 Set | 93988_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423567_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423568_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423567_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423568_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423568_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423568_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423567_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423567_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 320617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2230131 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2630328 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2630328 | - | - |
| mouse-6 v1.1 (Illumina) | scl18232.3.7_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230131 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18232.3.7_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230131 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2793488 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630328 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630328 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2793488 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18232.3.7_30-S | - | - |

Psma7
Genomic Map