Assay Details
| Gene Symbol: Add2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271857.1 | NP_001258786.1 | 6 - 7 | 959 | 58 | |
| NM_001271858.1 | NP_001258787.1 | 6 - 7 | 956 | 58 | ||
| NM_001271859.1 | NP_001258788.1 | 6 - 7 | 757 | 58 | ||
| NM_001271860.1 | NP_001258789.1 | 6 - 7 | 959 | 58 | ||
| NM_001271861.1 | NP_001258790.1 | 6 - 7 | 760 | 58 | ||
| NM_013458.5 | NP_038486.2 | 6 - 7 | 760 | 58 | ||
| GenBank mRNA | AF100422.1 | - | 5 - 6 | 635 | 58 | |
| AF100423.1 | - | 5 - 6 | 635 | 58 | ||
| AF189769.1 | - | 4 - 5 | 557 | 58 | ||
| AF189770.1 | - | 4 - 5 | 557 | 58 | ||
| AK014496.1 | - | 6 - 7 | 725 | 58 | ||
| AK028425.1 | - | 6 - 7 | 731 | 58 | ||
| AK029488.1 | - | 6 - 7 | 910 | 58 | ||
| AK147544.1 | - | 6 - 7 | 832 | 58 | ||
| AK156954.1 | - | 6 - 7 | 749 | 58 | ||
| AK160736.1 | - | 6 - 7 | 913 | 58 | ||
| BC046783.1 | - | 6 - 7 | 893 | 6401200 | 58 | |
| BC053032.1 | - | 6 - 7 | 893 | 6401200 | 58 | |
More Information
Set Membership: |
|

Add2
Genomic Map