Assay Details
| Gene Symbol: Anxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013471.2 | NP_038499.2 | 5 - 6 | 419 | 88 | |
| GenBank mRNA | AK032703.1 | - | 4 - 5 | 447 | 88 | |
| AK132293.1 | - | 6 - 7 | 461 | 88 | ||
| AK150486.1 | - | 5 - 6 | 380 | 88 | ||
| AK150614.1 | - | 5 - 6 | 366 | 88 | ||
| AK151054.1 | - | 5 - 6 | 366 | 88 | ||
| AK151236.1 | - | 5 - 6 | 398 | 88 | ||
| AK167338.1 | - | 5 - 6 | 420 | 88 | ||
| AK168390.1 | - | 5 - 6 | 391 | 88 | ||
| AK168487.1 | - | 5 - 6 | 400 | 88 | ||
| AK168917.1 | - | 4 - 5 | 424 | 88 | ||
| AK170447.1 | - | 5 - 6 | 365 | 88 | ||
| BC055871.1 | - | 5 - 6 | 379 | 4947415 | 88 | |
| U72941.1 | - | 5 - 6 | 390 | 88 | ||
| U95371.1 | - | 5 - 6 | 390 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa4
Genomic Map