| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100013175.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013175.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013175.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013186.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013186.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013186.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013187.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013187.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013187.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013178.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013178.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013178.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013179.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013179.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013179.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013183.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013183.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013183.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013176.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013176.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013176.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013182.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013182.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013182.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013188.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013188.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013188.mm.1 |
TC0100001595.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013177.mm.1 |
NM_018729.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013177.mm.1 |
NM_018729.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013177.mm.1 |
TC0100001595.mm.1 |
MTA TC |