Assay Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019800.4 | NP_062774.2 | 5 - 6 | 1012 | 55 | |
| GenBank mRNA | AB030038.1 | - | 5 - 6 | 1005 | 55 | |
| AF216223.1 | - | 5 - 6 | 489 | 55 | ||
| AK002783.1 | - | 5 - 6 | 926 | 55 | ||
| AK077729.1 | - | 5 - 6 | 1000 | 55 | ||
| AK160271.1 | - | 5 - 6 | 931 | 55 | ||
| AK165356.1 | - | 5 - 6 | 1013 | 55 | ||
| AK171757.1 | - | 5 - 6 | 927 | 55 | ||
| BC047276.1 | - | 5 - 6 | 752 | 6487551 | 55 | |
More Information
Set Membership: |
|

Acp6
Genomic Map