Assay Details
| Gene Symbol: Mllt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019914.4 | NP_064298.1 | 1 - 2 | 593 | 77 | |
| GenBank mRNA | AB000733.1 | - | 1 - 2 | 167 | 77 | |
| AK046005.1 | - | 1 - 2 | 98 | 77 | ||
| AK152708.1 | - | 1 - 2 | 593 | 77 | ||
| AK158495.1 | - | 1 - 2 | 219 | 77 | ||
| BC022963.1 | - | 1 - 2 | 34 | 5365628 | 77 | |
| U95498.1 | - | 1 - 2 | 154 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB000733.1,AK046005.1,AK152708.1,AK158495.1,BC022963.1,NM_019914.4,U95498.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of mitochondrial depolarization
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
intrinsic apoptotic signaling pathway
molecular_function
positive regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of mitochondrial depolarization
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
intrinsic apoptotic signaling pathway
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342936 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469731 | BC028768.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273751 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65106 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124113 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128377 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | U95498.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | U95498.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342918 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00702247 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128377 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124113 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04994542 | - | - |
| Whole Mouse Genome G4122A | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | BC022963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | U95498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK046005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK152708.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P379660 | AK158495.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116454 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124113 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613423 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613414 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02613420 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02613425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02957982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02860217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02860226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02613423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02570417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02647277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02613413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02950775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03145910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02385241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500116 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416313_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416314_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500114 | - | - |
| Mouse Genome 430 2.0 Array | 1416313_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416313_at | - | - |
| Mouse Genome 430 2.0 Array | 1416314_at | - | - |
| Mouse Genome 430 2.0 Array | 1416314_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56772_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56772_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019092.mm.1 | NM_019914.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019092.mm.1 | NM_019914.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019092.mm.1 | TC0300002470.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019091.mm.1 | NM_019914.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019091.mm.1 | NM_019914.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019091.mm.1 | TC0300002470.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019094.mm.1 | NM_019914.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019094.mm.1 | NM_019914.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019094.mm.1 | TC0300002470.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019093.mm.1 | NM_019914.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019093.mm.1 | NM_019914.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019093.mm.1 | TC0300002470.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416314_at | - | - |
| Mouse Expression Set 430 | 1416313_PM_at | - | - |
| Mouse Expression Set 430 | 1416313_at | - | - |
| Mouse Expression Set 430 | 1416314_PM_at | - | - |
| Murine Genome U74v2 Set | 95503_at | - | - |
| Murine Genome U74v2 Set | 95503_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416313_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416314_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416313_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416314_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416314_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416313_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416313_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416314_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 514823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5080278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221429 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221429 | - | - |
| mouse-6 v1.1 (Illumina) | scl056772.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056772.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2988212 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221429 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221429 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2988212 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056772.1_0-S | - | - |

Mllt11
Genomic Map