Assay Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019977.2 | NP_064361.2 | 3 - 4 | 208 | 85 | |
| GenBank mRNA | AF197127.1 | - | 3 - 4 | 206 | 85 | |
| AK002406.1 | - | 3 - 4 | 222 | 85 | ||
| AY738257.1 | - | 2 - 3 | 178 | 85 | ||
| BC013543.1 | - | 3 - 4 | 208 | 4242159 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Miox
Genomic Map