Assay Details
| Gene Symbol: Prl2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019991.1 | NP_064375.1 | 3 - 4 | 365 | 81 | |
| GenBank mRNA | AB032428.1 | - | 3 - 4 | 386 | 81 | |
| AF226610.1 | - | 3 - 4 | 321 | 81 | ||
| AF234636.1 | - | 3 - 4 | 365 | 81 | ||
| AF272368.1 | - | 3 - 4 | 317 | 81 | ||
| AK005424.1 | - | 3 - 4 | 342 | 81 | ||
| BC099458.1 | - | 3 - 4 | 361 | 30790018 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl2a1
Genomic Map