Assay Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019993.3 | NP_064377.2 | 3 - 4 | 702 | 71 | |
| GenBank mRNA | AB296386.1 | - | 3 - 4 | 647 | 71 | |
| AF170919.1 | - | 3 - 4 | 447 | 71 | ||
| AK132336.1 | - | 3 - 4 | 487 | 71 | ||
| AK149589.1 | - | 3 - 4 | 488 | 71 | ||
| AK150093.1 | - | 3 - 4 | 445 | 71 | ||
| AK154914.1 | - | 3 - 4 | 686 | 71 | ||
| AK162354.1 | - | 3 - 4 | 486 | 71 | ||
| AK165730.1 | - | 3 - 4 | 702 | 71 | ||
| AK168261.1 | - | 3 - 4 | 71 | |||
| AK168309.1 | - | 3 - 4 | 71 | |||
| AK168878.1 | - | 3 - 4 | 651 | 71 | ||
| BC003297.1 | - | 3 - 4 | 479 | 3583976 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh9a1
Genomic Map