Assay Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019993.3 | NP_064377.2 | 6 - 7 | 1174 | 62 | |
| GenBank mRNA | AB296386.1 | - | 6 - 7 | 1119 | 62 | |
| AF170919.1 | - | 6 - 7 | 919 | 62 | ||
| AK132336.1 | - | 6 - 7 | 959 | 62 | ||
| AK149589.1 | - | 6 - 7 | 960 | 62 | ||
| AK150093.1 | - | 6 - 7 | 917 | 62 | ||
| AK154914.1 | - | 6 - 7 | 1158 | 62 | ||
| AK162354.1 | - | 6 - 7 | 958 | 62 | ||
| AK168261.1 | - | 6 - 7 | 1152 | 62 | ||
| AK168309.1 | - | 6 - 7 | 1152 | 62 | ||
| AK168878.1 | - | 6 - 7 | 1123 | 62 | ||
| AK180861.1 | - | 1 - 2 | 125 | 62 | ||
| AK188805.1 | - | 1 - 2 | 125 | 62 | ||
| AK199575.1 | - | 1 - 2 | 125 | 62 | ||
| AK203750.1 | - | 1 - 2 | 125 | 62 | ||
| AK211951.1 | - | 1 - 2 | 125 | 62 | ||
| BC003297.1 | - | 6 - 7 | 951 | 3583976 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh9a1
Genomic Map