Assay Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB038560.1 | - | 6 - 7 | 3335 | 99 | |
| AK050819.1 | - | 7 - 8 | 2676 | 99 | ||
| AK220315.1 | - | 6 - 7 | 3283 | 99 | ||
| BC025857.1 | - | 1 - 2 | 816 | 99 | ||
| BC092394.1 | - | 6 - 7 | 3353 | 30534262 | 99 | |
| BC141058.1 | - | 6 - 7 | 3291 | 9055670 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigo
Genomic Map