Assay Details
| Gene Symbol: Igdcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290315.1 | NP_001277244.1 | 15 - 16 | 2757 | 61 | |
| NM_020043.3 | NP_064427.2 | 15 - 16 | 2754 | 61 | ||
| GenBank mRNA | AB052620.1 | - | 15 - 16 | 2754 | 61 | |
| AB052621.1 | - | 15 - 16 | 2757 | 61 | ||
| AF176694.1 | - | 15 - 16 | 2705 | 61 | ||
| AK122535.1 | - | 2833 | 61 | |||
| CB524810.1 | - | 2 - 3 | 216 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc4
Genomic Map