Assay Details
| Gene Symbol: Hs1bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021429.3 | NP_067404.2 | 5 - 6 | 829 | 91 | |
| GenBank mRNA | AJ132192.1 | - | 5 - 6 | 823 | 91 | |
| AK040654.1 | - | 5 - 6 | 830 | 91 | ||
| AK043263.1 | - | 1 - 2 | 79 | 91 | ||
| AK084802.1 | - | 5 - 6 | 820 | 91 | ||
| AK154696.1 | - | 5 - 6 | 825 | 91 | ||
| AK170838.1 | - | 5 - 6 | 816 | 91 | ||
| BC095977.1 | - | 5 - 6 | 826 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hs1bp3
Genomic Map