Assay Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290764.1 | NP_001277693.1 | 13 - 14 | 1354 | 69 | |
| NM_001290766.1 | NP_001277695.1 | 11 - 12 | 1338 | 69 | ||
| NM_001290767.1 | NP_001277696.1 | 11 - 12 | 1109 | 69 | ||
| NM_021549.3 | NP_067524.2 | 12 - 13 | 1446 | 69 | ||
| GenBank mRNA | AF129451.1 | - | 11 - 12 | 1104 | 69 | |
| AK049992.1 | - | 12 - 13 | 1249 | 69 | ||
| AK159535.1 | - | 12 - 13 | 1211 | 69 | ||
| AK159770.1 | - | 12 - 13 | 1211 | 69 | ||
| AK160020.1 | - | 12 - 13 | 1211 | 69 | ||
| AK160160.1 | - | 13 - 14 | 1351 | 69 | ||
| AK172558.1 | - | 11 - 12 | 1143 | 69 | ||
| BC047269.1 | - | 10 - 11 | 1388 | 69 | ||
| BC057659.1 | - | 12 - 13 | 1248 | 2865792 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnkp
Genomic Map