Assay Details
| Gene Symbol: Ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021710.3 | NP_068356.1 | 1 - 2 | 345 | 153 | |
| GenBank mRNA | AF092093.1 | - | 1 - 2 | 274 | 153 | |
| AK005283.1 | - | 1 - 2 | 95 | 153 | ||
| BC001985.1 | - | 1 - 2 | 345 | 3495981 | 153 | |
| BC053339.1 | - | 1 - 2 | 105 | 5370721 | 153 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF092093.1,AK005283.1,BC001985.1,BC053339.1,NM_021710.3 transcripts.
Set Membership: |
|

Ap4s1
Genomic Map