Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023270.5 | NP_075759.3 | 1 - 2 | 668 | 106 | |
| GenBank mRNA | AB041548.1 | - | 1 - 2 | 668 | 106 | |
| AF426411.1 | - | 1 - 2 | 482 | 106 | ||
| AK004847.1 | - | 1 - 2 | 669 | 106 | ||
| AK008312.1 | - | 1 - 2 | 612 | 106 | ||
| AK018582.1 | - | 1 - 2 | 670 | 106 | ||
| AK146266.1 | - | 1 - 2 | 665 | 106 | ||
| AK167031.1 | - | 1 - 2 | 654 | 106 | ||
| AY112656.1 | - | 1 - 2 | 655 | 106 | ||
| BC010477.1 | - | 1 - 2 | 657 | 3154433 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf128
Genomic Map