Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254761.1 | NP_001241690.1 | 3 - 4 | 865 | 63 | |
| NM_023270.5 | NP_075759.3 | 3 - 4 | 991 | 63 | ||
| GenBank mRNA | AB041548.1 | - | 3 - 4 | 991 | 63 | |
| AF426411.1 | - | 3 - 4 | 805 | 63 | ||
| AK004847.1 | - | 3 - 4 | 992 | 63 | ||
| AK018582.1 | - | 3 - 4 | 993 | 63 | ||
| AK146266.1 | - | 3 - 4 | 988 | 63 | ||
| AK167031.1 | - | 3 - 4 | 977 | 63 | ||
| AY112656.1 | - | 3 - 4 | 978 | 63 | ||
| BC010477.1 | - | 3 - 4 | 980 | 3154433 | 63 | |
| DV064262.1 | - | 3 - 4 | 628 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf128
Genomic Map