Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254761.1 | NP_001241690.1 | 5 - 6 | 1045 | 106 | |
| NM_023270.5 | NP_075759.3 | 5 - 6 | 1171 | 106 | ||
| GenBank mRNA | AB041548.1 | - | 5 - 6 | 1171 | 106 | |
| AF426411.1 | - | 5 - 6 | 985 | 106 | ||
| AK004847.1 | - | 5 - 6 | 1172 | 106 | ||
| AK018582.1 | - | 5 - 6 | 1173 | 106 | ||
| AK167031.1 | - | 5 - 6 | 1157 | 106 | ||
| AY112656.1 | - | 5 - 6 | 1158 | 106 | ||
| BC010477.1 | - | 5 - 6 | 1160 | 3154433 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf128
Genomic Map