Assay Details
| Gene Symbol: Fip1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159573.1 | NP_001153045.1 | 6 - 7 | 593 | 96 | |
| NM_001159574.1 | NP_001153046.1 | 6 - 7 | 593 | 96 | ||
| NM_024183.5 | NP_077145.2 | 6 - 7 | 593 | 96 | ||
| GenBank mRNA | AK005061.1 | - | 6 - 7 | 562 | 96 | |
| AK008561.1 | - | 6 - 7 | 565 | 96 | ||
| AK049672.1 | - | 6 - 7 | 579 | 96 | ||
| AK164105.1 | - | 6 - 7 | 559 | 96 | ||
| AK168464.1 | - | 6 - 7 | 594 | 96 | ||
| BC003263.1 | - | 6 - 7 | 551 | 3499130 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fip1l1
Genomic Map