Assay Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289524.1 | NP_001276453.1 | 9 - 10 | 1150 | 129 | |
| NM_001289525.1 | NP_001276454.1 | 7 - 8 | 897 | 129 | ||
| NM_024194.6 | NP_077156.2 | 9 - 10 | 1153 | 129 | ||
| GenBank mRNA | AK011726.1 | - | 9 - 10 | 1134 | 129 | |
| AK017814.1 | - | 9 - 10 | 1122 | 129 | ||
| AK034978.1 | - | 1133 | 129 | |||
| AK148748.1 | - | 9 - 10 | 1113 | 129 | ||
| AK152244.1 | - | 7 - 8 | 878 | 129 | ||
| AK163288.1 | - | 9 - 10 | 1121 | 129 | ||
| AK170052.1 | - | 9 - 10 | 1130 | 129 | ||
| BC002279.1 | - | 5 - 6 | 542 | 3497400 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc40
Genomic Map