Assay Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025358.3 | NP_079634.2 | 8 - 9 | 838 | 70 | |
| GenBank mRNA | AK003137.1 | - | 8 - 9 | 820 | 70 | |
| AK003214.1 | - | 8 - 9 | 811 | 70 | ||
| AK012366.1 | - | 8 - 9 | 818 | 70 | ||
| BC005760.1 | - | 7 - 8 | 719 | 70 | ||
| BC058378.1 | - | 8 - 9 | 806 | 6820787 | 70 | |
More Information
Set Membership: |
|

Ndufa9
Genomic Map