Assay Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025358.3 | NP_079634.2 | 9 - 10 | 929 | 66 | |
| GenBank mRNA | AK003137.1 | - | 9 - 10 | 911 | 66 | |
| AK003214.1 | - | 9 - 10 | 902 | 66 | ||
| AK012366.1 | - | 9 - 10 | 909 | 66 | ||
| BC005760.1 | - | 8 - 9 | 810 | 66 | ||
| BC058378.1 | - | 9 - 10 | 897 | 6820787 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufa9
Genomic Map