Assay Details
| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025384.3 | NP_079660.1 | 4 - 5 | 324 | 111 | |
| GenBank mRNA | AK002598.1 | - | 4 - 5 | 320 | 111 | |
| AK003396.1 | - | 4 - 5 | 334 | 111 | ||
| AK007806.1 | - | 4 - 5 | 313 | 111 | ||
| AK007833.1 | - | 4 - 5 | 313 | 111 | ||
| BC027509.1 | - | 4 - 5 | 324 | 1364626 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc15
Genomic Map