Assay Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025486.2 | NP_079762.1 | 2 - 3 | 202 | 111 | |
| GenBank mRNA | AK002499.1 | - | 2 - 3 | 190 | 111 | |
| AK005662.1 | - | 2 - 3 | 186 | 111 | ||
| AK131833.1 | - | 2 - 3 | 171 | 111 | ||
| BC005665.1 | - | 2 - 3 | 157 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem208
Genomic Map