Assay Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025486.2 | NP_079762.1 | 3 - 4 | 269 | 80 | |
| GenBank mRNA | AK002499.1 | - | 3 - 4 | 257 | 80 | |
| AK005662.1 | - | 3 - 4 | 253 | 80 | ||
| AK131833.1 | - | 3 - 4 | 238 | 80 | ||
| BC005665.1 | - | 3 - 4 | 224 | 80 | ||
| BC024608.1 | - | 3 - 4 | 220 | 4242513 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem208
Genomic Map